Key Documents
BR16520
BRAND® burette clamp
Zinc alloy, nickel-plated for 2 burettes, clamp PVC-coated
Synonym(s):
Accessories, Burette clamp, Burettes
About This Item
UNSPSC Code:
41122413
NACRES:
NB.01
packaging
pack of 1 ea
manufacturer/tradename
BRAND 16520
General description
Burettes up to 50 ml can be mounted in seconds. PVC-coated clamps provide a secure grip. Comfortable height adjustment. Graduations remain visible. Very sturdy design, easy to operate. Integral support connector.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)