MilliporeSigma
All Photos(4)
Documents
BR152810
BRAND® tubing connector, T-shape
polypropylene, for tubing i.d., 8 ‑ 9 mm
Synonym(s):
Tubing, Tubing Connectors
About This Item
UNSPSC Code:
40183110
NACRES:
NB.43
material
polypropylene
feature
for tubing i.d. 8 ‑ 9 mm
packaging
pack of 20 ea
manufacturer/tradename
BRAND 152810
Related Categories
General description
The BRAND® T-shaped tubing connector is constructed from translucent polypropylene (PP), allowing for easy visibility of fluid flow. This connector is designed to create a branching point in tubing systems, enabling the connection of multiple tubes at different angles. Its durable construction ensures a secure and reliable fit, minimizing the risk of leaks or disconnections during operation.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG