Documents
BR130254
BRAND® amber narrow-mouth bottle, LDPE, with screw cap
capacity 500 mL
Synonym(s):
Bottles, Plastics
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
bottle
low-density polyethylene cap
Joint
Joints threaded neck (GL25)
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 130254
capacity
500 mL
diam. × H
75 mm × 180 mm
General description
For storage of light-sensitive substances. With screw cap
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG