BR114940
BRAND® mini cooler
PC, for 12 tubes
Synonym(s):
Accessories, Microcentrifuge Tubes, Mini cooler, Sample Storage, Tubes
All Photos(1)
About This Item
Recommended Products
material
PC
description
bench temp.maintained -70°C time 45 min.
feature
for 12 tubes
manufacturer/tradename
BRAND 114940
General description
PC. Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench. Durable polycarbonate filled with non-toxic gel. Mini coolers hold twelve 0.5 ml to 2.0 ml tubes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG