Documents
BR112805
BRAND® forcep with pointed ends
PMP, L 115 mm
Synonym(s):
Forceps
About This Item
UNSPSC Code:
41122405
NACRES:
NB.35
material
PMP
feature
autoclavable at 121 C
manufacturer/tradename
BRAND 112805
L
115 mm
General description
The BRAND® Forceps with Pointed Ends are precision tools designed for handling delicate materials in laboratory settings. Made from high-quality PMP (polymethylpentene), these forceps are lightweight yet durable, making them ideal for various applications. Designed to withstand autoclaving at 121 °C, ensuring effective sterilization for safe use in sterile environments.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG