Key Documents
BMSMW960012CM
Benchmark 12Channel Manifold
12-channel, manifold
Select a Size
1 EACH
$502.00
$502.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
$502.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.01
$502.00
Please contact Customer Service for Availability
feature
12-channel manifold
manufacturer/tradename
Benchmark Scientific MW9600-12CM
General description
12 channel manifold for Accuris SmartWasher 96
Application
Washing microplates for ELISA assays
Choose from one of the most recent versions:
Certificates of Analysis (COA)