Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BMSE1200EUK

Benchmark myGel InstaView Gel Tank with Blue LED Illuminator

AC/DC input 230 V, UK plug

Synonym(s):

protein separation, gel imaging, gel casting, transillumination

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,060.00

$1,060.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$1,060.00

About This Item

UNSPSC Code:
41105300
NACRES:
NB.13

$1,060.00


Please contact Customer Service for Availability

manufacturer/tradename

Benchmark Scientific E1200-E-UK

Looking for similar products? Visit Product Comparison Guide

General description

myGel InstaView Gel Tank allows users to view sample migration during electrophoresis. Includes gel tank, casting set, blue LED transilluminator, orange filter cover and imaging enclosure. Compatible with any basic power supply

Application

Electrophoresis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service