Skip to Content
MilliporeSigma
All Photos(1)

Documents

BMSDP01503

Benchmark Pulse 150 Horn

diam. 3 mm

Synonym(s):

ultrasonic homogenizer, sonicator, pulsating sonication

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41101520
NACRES:
NB.13

material

titanium (allloy)

manufacturer/tradename

Benchmark Scientific DP0150-3

diam.

3 mm

compatibility

Benchmark Pulse 150 Ultrasonic Homogenizer

Looking for similar products? Visit Product Comparison Guide

Application

Ultrasonic homogenization and lysis

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service