Key Documents
BAH195590000
SP Bel-Art® Straight Tubing Connectors
tubing I.D. 0.5-0.5 in. (4.7-12.7 mm)
Synonym(s):
connectors, tubing, tubing coupler
Select a Size
12 EACH
$88.70
$88.70
Please contact Customer Service for Availability
Select a Size
Change View
12 EACH
$88.70
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
$88.70
Please contact Customer Service for Availability
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art H19559-0000
L × W
3.375 in. (85.7  mm) × 0.8125 in. (20.6 mm)
tubing I.D.
0.5-0.5 in. (4.7-12.7 mm)
General description
These straight fittings ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.).
Features and Benefits
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases.
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. does this item contain several different adaptors with different diameters or just on etype
    1 answer
    1. This product includes 12 tubing connectors of the same size as shown below.
      Helpful?