Documents
BAH131821015
Clavies® autoclave bags
width × H × thickness 10 in. (250 mm) × 15 in. (380 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Synonym(s):
sterilzation bag, transparent autoclave bags
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
clear
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art H13182-1015
width × H × thickness
10 in. (250 mm) × 15 in. (380 mm) × 2 mil (0.05 mm)
General description
These transparent autoclave bags are made of 0.05mm (2 mil) thick polypropylene for discarding used Petri dishes, membrane filters, multi-well cell culture plates, cell culture flasks, culture plates, pipettes, and more.
Features and Benefits
  • Three largest sizes hold 65, 200 and 240 standard Petri dishes respectively
  • Steam autoclavable at 135°C (275°F)
Legal Information
Clavies is a registered trademark of SP Scienceware, a division of SP