Key Documents
BAF420751002
SP Bel-Art® Secador® Mini Gas-Purge Desiccator Cabinet
polystyrene, volume 0.3 ft3
Select a Size
1 EACH
$486.00
$486.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
$486.00
About This Item
UNSPSC Code:
41123002
NACRES:
NB.77
$486.00
Please contact Customer Service for Availability
material
clear
polystyrene
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F42075-1002
volume
0.3 ft3
General description
The sturdy construction and compact size of this desiccator cabinet assures reliable storage for moisture sensitive products when benchtop space is at a premium.
  • Large side swinging door maximizes access to interior space; has a patented seal design with tab latches and security loop for padlock or tamper-evident seals; opens right to left
  • Top mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D.
  • Door mounted dial hygrometer indicates relative humidity at a glance
  • Clear polystyrene for easy viewing of contents
  • Gas ports can be closed to use desiccator with standard desiccant cartridges
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)