BAF378400000
Sampling spatula, polypropylene
non-sterile
All Photos(1)
About This Item
Recommended Products
sterility
non-sterile
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F37840-0000
L × W × H
152 mm × 19 mm × 13 mm
Overall L
9 in.
General description
One-piece spatula is molded from polypropylene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums.
- V-shaped spatula is perfect for sampling granulated or semi solid material
- Non-sterile
- Use different spatula for different materials to reduce cross contamination
- Sharp point will pierce most packaging