Documents
BAF369430020
SP Bel-Art® Sterileware Volumetric Sampling Spoons
white high-density polyethylene, capacity × L 20 mL (0.68 oz) × 18 cm (7.125 in.), sterile, pack of 100 ea (individually wrapped)
Synonym(s):
Lab Scoop, measuring spoon, spoon
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
white high-density polyethylene
sterility
sterile
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
Bel-Art F36943-0020
bowl diam.
1.75 in. (43 mm)
capacity × L
20 mL (0.68 oz) × 18 cm (7.125 in.)
General description
Suitable for use with powders, granules, liquids, gels or suspensions, these sterile spoons allow set volumes to be accurately measured and sampled.
Features and Benefits
  • Leveling to top of bowl provides accurately measured samples
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) spoons heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP