Documents
BAF203071825
SP Bel-Art® Safe-Lab Therm-O-Vac Joint Adapter
for for 24/40 Tapered Joints
Synonym(s):
thermovac adapter
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F20307-1825
hole diam.
6 mm
fitting
for for 24/40 Tapered Joints
General description
PTFE® adapter with Viton® O-ring on the body, screw cap and hose connection.
Features and Benefits
Not autoclavable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company