Key Documents
BAF183070003
Scienceware® Vikem® coated closed lead ring
for fits flask 5-10 mL
Select a Size
1 EACH
$43.90
$43.90
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
$43.90
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
$43.90
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18307-0003
fitting
for fits flask 5-10 mL
General description
Protect and stabilize expensive glassware with these Vikem® vinyl coated lead rings.
Features and Benefits
  • Stabilize glassware immersed in water baths;reduces floating, breakage and chemical spills
  • Bright red Vikem® vinyl coating
  • Chemical-resistant
  • Will not scratch flasks
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)