Documents
BAF169570000
SP Bel-Art® Cone Style Acid/Solvent Bottle Carrier
to hold, one 2.5 Liter Bottle (5 pints)
Synonym(s):
solvent bottle carrier
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
conical
feature
to hold one 2.5 Liter Bottle (5 pints)
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F16957-0000
diam. × H
7.125 in. (180 mm) × 13.5 in. (350 mm)
General description
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles.
Features and Benefits
  • Cone style lid has a 16mm (⅝”) opening in the top for insertion of tubing; simplifies the transfer of liquids without pouring
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard US reagent bottles
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP