Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAF131920002

SP Bel-Art® Biohazard disposal stand

to hold, 24 x 30 in (61 x 76 cm) bag

Synonym(s):

biohaz waste bag, biohazard disposal bags, waste bag

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$116.00

$116.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$116.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

$116.00


Please contact Customer Service for Availability

feature

to hold 24 x 30 in (61 x 76 cm) bag

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F13192-0002

H

21.5 in. (546 mm)
215 mm

Looking for similar products? Visit Product Comparison Guide

General description

Keep bags open and securely supported on these economical, lightweight, stainless steel wire frames. Polypropylene bottom serves as a containment tray in the event of bag leakage and is safety orange in color for easy identification as a biohazard containment system.

Features and Benefits

  • Small bag holder is ideal for benchtop use
  • Assembles in minutes and is permanent
  • Dishwasher-safe; autoclavable at 121°C (250°F)
  • Use F13192-0102 reusable, sanitary cover with medium and large Clavies Biohazard Bag Holders

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service