Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAF131660001

Scienceware® biohazard disposal bags

W × H × thickness 8 1/2 in. (22 cm) × 11 in. (28 cm) × 0.018 mm, orange-red, pack of 1000

Synonym(s):

autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EA
$315.00

$315.00


Please contact Customer Service for Availability


Select a Size

Change View
1000 EA
$315.00

About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

$315.00


Please contact Customer Service for Availability

material

orange-red

packaging

pack of 1000

manufacturer/tradename

Bel-Art F13166-0001

W × H × thickness

8 1/2 in. (22 cm) × 11 in. (28 cm) × 0.018 mm

Looking for similar products? Visit Product Comparison Guide

Application

Scienceware® biohazard disposal bags are suitable for disposing of used tips and tubes.

Features and Benefits

  • Contains one long flap for easy opening
  • Printed with the biohazard symbol and sterilization indicator patch
  • Made of high molecular weight, high-density (HMHD) polyethylene
  • The small size can easily fit over your hand when inverted and ensures a safe pick-up of waste.
  • Steam autoclavable at 121°C (250°F)

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service