BAF116466250
Scienceware® safety labeled wash bottles
label, Dichloromethane, wide-mouth yellow cap, capacity 500 mL (16 oz)
Synonym(s):
dispensing bottle, safety bottles, squeeze bottle
Select a Size
$93.60
Select a Size
About This Item
$93.60
Recommended Products
material
low-density polyethylene
wide-mouth yellow cap
feature
label Dichloromethane
packaging
pack of 6 ea
manufacturer/tradename
Bel-Art F11646-6250
capacity
500 mL
500 mL (16 oz)
color
opaque
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Easy-to-read printed bottles
- Wide mouth 53mm (2") bottle for easy refilling
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Polypropylene closures
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- Easy-to-read printed bottles
- Wide mouth 53mm (2") bottle for easy refilling
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Polypropylene closures
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service