Key Documents
BAB614007100
SP Bel-Art® H-B Beaker Clip Liquid-in-Glass Thermometer Holder
stainless steel, holds 4 thermometers
Synonym(s):
thermometer holder
About This Item
UNSPSC Code:
41122800
NACRES:
NB.79
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B61400-7100
width × L
38 mm (1.4 in.) × 3 in. (76 mm)
General description
Holds up to 4 thermometer probes with 3.5-4.0mm O.D. at one time.
Features and Benefits
Holds up to 4 thermometer probes with 3.5-4.0mm O.D. at one time.
  • Clips easily onto the side of any beaker
  • Made from SUS304 stainless steel
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP