Key Documents
BAB609011300
SP Bel-Art® H-B Durac® Thermocouple Thermometer Probe
K-TYPE
Select a Size
1 EACH
$190.00
$190.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
$190.00
About This Item
UNSPSC Code:
41112219
NACRES:
NB.79
$190.00
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60901-1300
General description
For use with H-B DURAC Calibrated Thermocouple Thermometers or others that accept K or J type probes.
Features and Benefits
  • For temperature measurement of liquids, semi-solids, or air/gasses
  • Measure wide temperature ranges
  • 15.24cm (6”) stainless steel probe
  • B60901-1300 perfect for use in dewars
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)