Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B92001

Sigma-Aldrich

4-tert-Butylcyclohexanol, mixture of cis and trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H10OH
CAS Number:
Molecular Weight:
156.27
Beilstein:
1902277
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

110-115 °C/15 mmHg (lit.)

mp

62-70 °C (lit.)

SMILES string

CC(C)(C)C1CCC(O)CC1

InChI

1S/C10H20O/c1-10(2,3)8-4-6-9(11)7-5-8/h8-9,11H,4-7H2,1-3H3

InChI key

CCOQPGVQAWPUPE-UHFFFAOYSA-N

Application

4-tert-Butylcyclohexanol (mixture of cis and trans) can be used as a reactant to synthesize tris(4,4′-di-tert-butyl-2,2′-bipyridine)(trans-4-tert-butylcyclohexanolato)deca-μ-oxido-heptaoxidoheptavanadium oxide cluster complex by reacting with [V8O20(C18H24N2)4].[1] It can also be used as a reactant in competitive Oppenauer oxidation experiments in the presence of zeolite BEA as a stereoselective catalyst. Only cis-isomer is selectively converted to the corresponding ketone, whereas trans-isomer remains unchanged.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tris (4, 4?-di-tert-butyl-2, 2?-bipyridine)(trans-4-tert-butylcyclohexanolato) deca- ? -oxido-heptaoxidoheptavanadium acetonitrile monosolvate including another unknown solvent molecule
Kodama S, et al.
IUCrData, 5(4), x200449-x200449 (2020)
Stereoselective Meerwein-Ponndorf-Verley and Oppenauer reactions catalysed by zeolite BEA
Creyghton EJ, et al.
J. Mol. Catal. A: Chem., 115(3), 457-472 (1997)
Mireia Oromí-Farrús et al.
Journal of analytical methods in chemistry, 2012, 452949-452949 (2012-06-01)
The use of iodine as a catalyst and either acetic or trifluoroacetic acid as a derivatizing reagent for determining the enantiomeric composition of acyclic and cyclic aliphatic chiral alcohols was investigated. Optimal conditions were selected according to the molar ratio

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service