Skip to Content
MilliporeSigma
All Photos(1)

Documents

B91757

Sigma-Aldrich

tert-Butylcyclohexane

≥99%

Synonym(s):

2-Cyclohexyl-2-methylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)C(CH3)3
CAS Number:
Molecular Weight:
140.27
Beilstein:
1900578
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor pressure

5 mmHg ( 37.7 °C)

Assay

≥99%

refractive index

n20/D 1.447 (lit.)

bp

167 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C1CCCCC1

InChI

1S/C10H20/c1-10(2,3)9-7-5-4-6-8-9/h9H,4-8H2,1-3H3

InChI key

XTVMZZBLCLWBPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service