Skip to Content
MilliporeSigma
All Photos(2)

Documents

B90408

Sigma-Aldrich

sec-Butylbenzene

≥99%

Synonym(s):

2-Phenylbutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)CH2CH3
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903902
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

4 mmHg ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

784 °F

refractive index

n20/D 1.489 (lit.)

bp

173-174 °C (lit.)

mp

−75.5 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

CCC(C)c1ccccc1

InChI

1S/C10H14/c1-3-9(2)10-7-5-4-6-8-10/h4-9H,3H2,1-2H3

InChI key

ZJMWRROPUADPEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewa Kaczorek et al.
Chemosphere, 90(2), 471-478 (2012-08-29)
Pseudomonas stutzeri strain 9 was isolated from petroleum-contaminated soil. The main purpose of this study was to investigate how the long-term contact of this strain with diesel oil influences its surface and biodegradation properties. The experiments showed that the tested
Metabolites of three structural isomers of butylbenzene in the bile of rainbow trout.
J Hellou et al.
Bulletin of environmental contamination and toxicology, 44(3), 487-493 (1990-03-01)
Qing Lv et al.
Journal of chromatography. A, 1404, 89-94 (2015-06-09)
Cyclotriveratrylene (CTV) is reported here for the first time as stationary phase for capillary gas chromatographic (GC) separations. CTV stationary phase showed weak polarity comparable to the conventional 5% phenyl polysiloxane stationary phase but exhibited different retention behaviours and higher
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service