Skip to Content
MilliporeSigma
All Photos(1)

Documents

B80003

Sigma-Aldrich

N-(3-Bromopropyl)phthalimide

98%

Synonym(s):

(γ-Bromopropyl)phthalimide, 1-Phthalimido-3-bromopropane, 2-(3-Bromopropyl)isoindole-1,3-dione, 3-(Phthalimido)propyl bromide, 3-Bromo-N-phthaloylpropylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10BrNO2
CAS Number:
Molecular Weight:
268.11
Beilstein:
157547
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

reaction suitability

reagent type: cross-linking reagent

mp

72-74 °C (lit.)

functional group

bromo

SMILES string

BrCCCN1C(C2=CC=CC=C2C1=O)=O

InChI

1S/C11H10BrNO2/c12-6-3-7-13-10(14)8-4-1-2-5-9(8)11(13)15/h1-2,4-5H,3,6-7H2

InChI key

VKJCJJYNVIYVQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service