Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B54800

Sigma-Aldrich

Tetrazolium Blue Chloride

Synonym(s):

3,3′-(3,3′-Dimethoxy[1,1′-biphenyl]-4,4′-diyl)-bis(2,5-diphenyl-2H-tetrazolium) dichloride, 3,3′-Dianisole-4,4′-bis(3,5-diphenyltetrazolium chloride), BTC, Blue Tetrazolium chloride, TZ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H32Cl2N8O2
CAS Number:
Molecular Weight:
727.64
Beilstein:
3894077
EC Number:
MDL number:
UNSPSC Code:
12171500

mp

255 °C (dec.) (lit.)

λmax

254 nm

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccccc3)-c4ccccc4)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccccc7)-c8ccccc8

InChI

1S/C40H32N8O2.2ClH/c1-49-37-27-31(23-25-35(37)47-43-39(29-15-7-3-8-16-29)41-45(47)33-19-11-5-12-20-33)32-24-26-36(38(28-32)50-2)48-44-40(30-17-9-4-10-18-30)42-46(48)34-21-13-6-14-22-34;;/h3-28H,1-2H3;2*1H/q+2;;/p-2

InChI key

MUUHXGOJWVMBDY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service