Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B5131

Sigma-Aldrich

6-Benzyloxyindole

crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

crystalline

color

yellow to brown

storage temp.

2-8°C

SMILES string

C(Oc1ccc2cc[nH]c2c1)c3ccccc3

InChI

1S/C15H13NO/c1-2-4-12(5-3-1)11-17-14-7-6-13-8-9-16-15(13)10-14/h1-10,16H,11H2

InChI key

FPMICYBCFBLGOZ-UHFFFAOYSA-N

Application

  • Reactant for enantioselective synthesis of quaternary carbon-containing 3-(3-indolyl)isoindolin-1-ones
  • Reactant for Friedel-Crafts alkylation reactions with hydroxyisoindolinones
  • Reactant for direct and regioselective synthesis of β-heteroarylated ketones
  • Reactant for preparation of hepatitis C virus (HCV) inhibitors
  • Reactant for preparation of protein kinase C (PKC) inhibitors
  • Reactant for preparation of indol-3-yl tetramethylcyclopropyl ketones as CB2 cannabinoid receptor ligands

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nenad Manevski et al.
PloS one, 8(1), e54767-e54767 (2013-02-02)
Human UDP-glucuronosyltransferases (UGTs) are important enzymes in metabolic elimination of endo- and xenobiotics. It was recently shown that addition of fatty acid free bovine serum albumin (BSA) significantly enhances in vitro activities of UGTs, a limiting factor in in vitro-in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service