Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

B47707

Sigma-Aldrich

4,4-Bis(4-hydroxyphenyl)valeric acid

95%

Synonym(s):

Diphenolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(C6H4OH)2CH2CH2CO2H
CAS Number:
Molecular Weight:
286.32
Beilstein:
1886361
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

167-170 °C (lit.)

SMILES string

CC(CCC(O)=O)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C17H18O4/c1-17(11-10-16(20)21,12-2-6-14(18)7-3-12)13-4-8-15(19)9-5-13/h2-9,18-19H,10-11H2,1H3,(H,20,21)

InChI key

VKOUCJUTMGHNOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldobenedetto Zotti et al.
Nanomaterials (Basel, Switzerland), 10(2) (2020-01-26)
The effects of the addition of an aromatic hyperbranched polyester (AHBP) on thermal, mechanical, and fracture toughness properties of a thermosetting resin system were investigated. AHBP filler, synthesized by using a bulk poly-condensation reaction, reveals a glassy state at room

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service