Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00627

Sigma-Aldrich

6-(2-(5-chloropyridin-2-yl)-1-phenylpropan-2-yl)pyridin-2-ol

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$264.00
1 G
$742.00

$264.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$264.00
1 G
$742.00

About This Item

Empirical Formula (Hill Notation):
C19H17ClN2O
Molecular Weight:
324.80
UNSPSC Code:
12352100
NACRES:
NA.21

$264.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

Quality Level

mp

188-189 °C

storage temp.

−20°C

Application

This ligand was developed by the Yu lab to enable the Pd-catalyzed dehydrogenation of aliphatic carboxylic acids through a typically challenging activation of the β-methylene C-H bond. This method is chemoselective to carboxylic acids even in the presence of other enolizable functional groups, and may also viably utilize molecular oxygen as the terminal oxidant for the transformation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Li et al.
Science (New York, N.Y.), 372(6549), 1452-1457 (2021-11-30)
Hydroxylation of aryl carbon-hydrogen bonds with transition metal catalysts has proven challenging when oxygen is used as the oxidant. Here, we report a palladium complex bearing a bidentate pyridine/pyridone ligand that efficiently catalyzes this reaction at ring positions adjacent to
Zhen Wang et al.
Science (New York, N.Y.), 374(6572), 1281-1285 (2021-11-12)
Dehydrogenative transformations of alkyl chains to alkenes through methylene carbon-hydrogen (C–H) activation remain a substantial challenge. We report two classes of pyridine-pyridone ligands that enable divergent dehydrogenation reactions through palladium-catalyzed β-methylene C–H activation of carboxylic acids, leading to the direct

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service