Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00608

Sigma-Aldrich

Ni(bpy)3Br2

Synonym(s):

Trisbipyridylnickel dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H24Br2N6Ni
Molecular Weight:
687.05
UNSPSC Code:
12352300

form

powder or crystals

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

Application

Ni(bpy)3Br2 is a Ni precatalyst shown by the Baran Lab to be an effective catalyst for the electrochemical amination (e-amination) of arly halides with amines.

Product can be used with our Synlectro electrochemistry platform (ESYNTH001)

related product

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1B - STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Kawamata et al.
Journal of the American Chemical Society, 141(15), 6392-6402 (2019-03-25)
C-N cross-coupling is one of the most valuable and widespread transformations in organic synthesis. Largely dominated by Pd- and Cu-based catalytic systems, it has proven to be a staple transformation for those in both academia and industry. The current study

Related Content

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service