Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00456

Sigma-Aldrich

Sodium N-benzyloxycarbonyl-4-piperidinesulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16NNaO4S
Molecular Weight:
305.33
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

reaction type: C-C Bond Formation
reaction type: C-H Activation
reagent type: catalyst

SMILES string

O=S(O[Na])C1CCN(C(OCC2=CC=CC=C2)=O)CC1

InChI

1S/C13H17NO4S.Na/c15-13(18-10-11-4-2-1-3-5-11)14-8-6-12(7-9-14)19(16)17;/h1-5,12H,6-10H2,(H,16,17);/q;+1/p-1

InChI key

LCXWFQBBQAGBHZ-UHFFFAOYSA-M

General description

Sodium N-benzyloxycarbonyl-4-piperidinesulfinate is a sulfinate reagent bearing an N-substituted piperidine moiety.

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service