Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00428

Sigma-Aldrich

4,4′-Dihydroxydiphenyl sulfone bisfluorosulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8F2O8S3
CAS Number:
Molecular Weight:
414.38
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

InChI

1S/C12H8F2O8S3/c13-24(17,18)21-9-1-5-11(6-2-9)23(15,16)12-7-3-10(4-8-12)22-25(14,19)20/h1-8H

InChI key

HLYHIXFVAKOJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service