Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00220

Sigma-Aldrich

2,3,4-Trifluorobenzenesulfonyl fluoride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2F4O2S
Molecular Weight:
214.14
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product ALD00220, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

95%

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.453

density

1.596 g/mL at 25 °C

SMILES string

FS(C1=C(F)C(F)=C(F)C=C1)(=O)=O

InChI

1S/C6H2F4O2S/c7-3-1-2-4(13(10,11)12)6(9)5(3)8/h1-2H

InChI key

QSTVOESYDQCSJJ-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

216.0 °F

Flash Point(C)

102.22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In collaboration with K. Barry Sharpless and coworkers, we offer a variety of sulfonyl fluoride reagents that undergo a “Click II” reaction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service