Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00106

Sigma-Aldrich

5-Methoxycarbonyl-1,2,3-triazine

Synonym(s):

1,2,3-Triazine-5-carboxylic acid, methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N3O2
CAS Number:
Molecular Weight:
139.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

storage temp.

−20°C

SMILES string

O=C(OC)C1=CN=NN=C1

InChI

1S/C5H5N3O2/c1-10-5(9)4-2-6-8-7-3-4/h2-3H,1H3

InChI key

DZONTPIFZFRSGP-UHFFFAOYSA-N

Application

1,2,3-Triazines have been shown to be reactive substrates in inverse electron demand Diels-Alder strategies. Recent examples by the Boger Research Group have utilized this reactive motif in the construction of highly functionalized N-containing heterocycles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Inverse electron demand Diels-Alder reactions enable total synthesis of natural products with heteroaromatic ring systems.

Related Content

Boger Lab utilizes nature's solutions in complex natural products, extending them through rational design for improved biological outcomes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service