Skip to Content
MilliporeSigma
All Photos(1)

Documents

ALD00032

Sigma-Aldrich

1-(Oxazolo[4,5-b]pyridin-2-yl)-6-phenylhexan-1-one

Synonym(s):

1-(Oxazolo[4,5-b]pyridin-2-yl)-1-oxo-6-phenylhexane, 1-Oxazolo[4,5-b]pyridin-2-yl-6-phenyl-1-hexanone, OL-100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O2
CAS Number:
Molecular Weight:
294.35
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

storage temp.

−20°C

SMILES string

O=C(C1=NC2=C(C=CC=N2)O1)CCCCCC3=CC=CC=C3

InChI

1S/2C18H18N2O2/c2*21-15(18-20-17-16(22-18)12-7-13-19-17)11-6-2-5-10-14-8-3-1-4-9-14/h2*1,3-4,7-9,12-13H,2,5-6,10-11H2

InChI key

LFAKEBRRAYWLPI-UHFFFAOYSA-N

Application

Inhibitor of Fatty acid amide hydrolase (FAAH); a serine hydrolase resposible for the transformation of anandamide to arachidonic acid

Exceptionally potent inhibitors of fatty acid amide hydrolase: The enzyme responsible for degradation of endogenous oleamide and anandamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

As the exploration of the properties of complex natural products becomes increasingly more sophisticated with the technological advances being made in their screening and evaluation and as structural details of their interaction with biological targets becomes more accessible, the importance and opportunities for providing unique solutions to complex biological problems has grown. The Boger Lab addresses these challenging problems by understanding the complex solutions and subtle design elements that nature has provided in the form of a natural product and work to extend the solution through rational design elements to provide more selective, more efficacious, or more potent agents designed specifically for the problem or target under investigation. The resulting efforts have reduced many difficult or intractable synthetic challenges to manageable problems providing an approach not only to the natural product but one capable of simple extrapolation to a series of structural analogs with improved selectivity and efficacy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service