Skip to Content
MilliporeSigma
All Photos(1)

Documents

A9584

Sigma-Aldrich

N-Acetoxy-N-acetyl-4-chlorobenzenesulfonamide

≥98%

Synonym(s):

O-Acetyl-N-(4-chlorophenylsulfonyl)acethydroxamic acid, HOPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10ClNO5S
CAS Number:
Molecular Weight:
291.71
Beilstein:
5821854
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98%

form

flakes

storage temp.

2-8°C

SMILES string

CC(=O)ON(C(C)=O)S(=O)(=O)c1ccc(Cl)cc1

InChI

1S/C10H10ClNO5S/c1-7(13)12(17-8(2)14)18(15,16)10-5-3-9(11)4-6-10/h3-6H,1-2H3

InChI key

GWYBSWWLKXEDLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nitroxyl (HNO) releasing compound in neutral solution

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Fukuto et al.
Biochemical and biophysical research communications, 187(3), 1367-1373 (1992-09-30)
N,O-Diacylated-N-hydroxyarylsulfonamides are capable of slowly releasing nitroxyl (HNO) by simple, non-enzymatic hydrolysis in Krebs solution at 37 degrees C. Release of nitric oxide (NO) was not seen. These compounds were also found to elicit vasorelaxation in rabbit thoracic aorta in
M J Lee et al.
Journal of medicinal chemistry, 35(20), 3648-3652 (1992-10-02)
In the preceding paper, analogs of chlorpropamide with an OMe substituent on the sulfonamide nitrogen were shown to inhibit aldehyde dehydrogenase (AlDH), and it was postulated that these compounds were bioactivated by O-demethylation to release nitroxyl (HN = O, nitrosyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service