Skip to Content
MilliporeSigma
All Photos(1)

Documents

A91205

Sigma-Aldrich

Anthrone

96%

Synonym(s):

9(10H)-Anthracenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O
CAS Number:
Molecular Weight:
194.23
Beilstein:
1910173
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Assay

96%

mp

154-157 °C (lit.)

SMILES string

O=C1c2ccccc2Cc3ccccc13

InChI

1S/C14H10O/c15-14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-8H,9H2

InChI key

RJGDLRCDCYRQOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anthrone (a tricyclic aromatic ketone) on mixing with sulfuric acid forms anthrone reagent, which is widely used in the estimation of sugars. Anthrone shows an inhibitory effect on the peroxidation of linoleic acid. It undergoes Michael addition with cinnamaldehyde in the presence of proline or prolinol derivatives.

Application

Anthrone has been used in colorimetric anthrone assay. It may be used to prepare azomethines by reacting with amines.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric organocatalytic anthrone additions to activated alkenes.
Zea A, et al.
Tetrahedron, 67(14), 2513-2529 (2011)
Synthesis of azomethines of anthrone.
Nadkarny VV and Sanzgiri MN.
J. Ind. Chem. Soc., 67(6), 513-514 (1990)
The estimation of carbohydrates in plant extracts by anthrone.
Yemm EW and Willis AJ.
The Biochemical Journal, 57(3), 508-508 (1954)
Antioxidant activity of anthraquinones and anthrone.
Yen GC, et al.
Food Chemistry, 70(4), 437-441 (2000)
Liselotte Krenn et al.
Chemical & pharmaceutical bulletin, 52(4), 391-393 (2004-04-02)
In a study of the anthraderivatives in roots of Rheum emodi, three new anthrone C-glucosides, named 10-hydroxycascaroside C (1), 10-hydroxycascaroside D (2) and 10R-chrysaloin 1-O-beta-D-glucopyranoside (3) were isolated besides the rare compounds cascaroside C (4), cascaroside D (5) and cassialoin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service