Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A7675

Sigma-Aldrich

1-Acetyl-6-aminoindoline

powder

Synonym(s):

1-(6-Amino-2,3-dihydro-indol-1-yl)ethanone, 1-Acetyl-6-amino-2,3-dihydroindole, 6-Amino-N-acetylindoline, NSC 87623

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O
CAS Number:
Molecular Weight:
176.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

color

light yellow to light brown

storage temp.

2-8°C

SMILES string

CC(=O)N1CCc2ccc(N)cc12

InChI

1S/C10H12N2O/c1-7(13)12-5-4-8-2-3-9(11)6-10(8)12/h2-3,6H,4-5,11H2,1H3

InChI key

LOZKZWIQDVEDCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Transient receptor potential cation channel V1 (TRPV1) antagonists
  • Inhibitor of the human immunodeficiency virus (HIV) envelope glycoprotein gp120 binding to the cluster of differentiation 4 (CD4) receptor
  • Antiproliferative agents
  • Serotonin 5-HT2C/2B receptor antagonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service