Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

A55357

Sigma-Aldrich

1-(2-Aminoethyl)pyrrolidine

98%

Synonym(s):

2-Pyrrolidinoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
Beilstein:
102978
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.468 (lit.)

bp

66-70 °C/23 mmHg (lit.)

density

0.901 g/mL at 25 °C (lit.)

SMILES string

NCCN1CCCC1

InChI

1S/C6H14N2/c7-3-6-8-4-1-2-5-8/h1-7H2

InChI key

WRXNJTBODVGDRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(2-Aminoethyl)pyrrolidine is a heterocyclic building block. Nickel(II) salts of 1-(2-aminoethyl)pyrrolidine have been synthesized and thermal studies have been reported. The isomeric complexes trans-[NiL2(NCS)2] (violet) and cis-[NiL2(NCS)2] (blue) [L=1-(2-aminoethyl)pyrrolidine] have been synthesized and their X-ray crystal structure have been studied.

Application

1-(2-Aminoethyl)pyrrolidine may be used in the preparation of one-dimensional coordination polymer of nickel(II), [(μN,S-NCS)2{Ni(ampy)}]n (ampy=1-(2-aminoethyl)pyrrolidine).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of cis and trans isomers of [NiL 2 (NCS) 2][L= 1-(2-aminoethyl) pyrrolidine]: X-ray single-crystal structures.
Laskar IR, et al.
Polyhedron, 19(15), 1803-1807 (2000)
Krzysztof Sztandera et al.
International journal of molecular sciences, 21(12) (2020-06-27)
Photodynamic therapy (PDT) is a skin cancer treatment alternative to chemotherapy and radiotherapy. This method exploits three elements: a phototoxic compound (photosensitizer), light source and oxygen. Upon irradiation by light of a specific wavelength, the photosensitizer generates reactive oxygen species
Syntheses, characterisation and solid state thermal studies of 1-(2-aminoethyl) piperidine (L), 1-(2-aminoethyl) pyrrolidine (L') and 4-(2-aminoethyl) morpholine (L ?) complexes of nickel (II):
Laskar IR, et al.
Polyhedron, 20(15), 2073-2082 (2001)
A 1D thiocyanato bridge nickel (II) system: crystal structure and magnetic property.
Maji TK, et al.
Polyhedron, 20(7), 651-655 (2001)
Juan Alfonso Redondo et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 117, 68-76 (2017-04-08)
The main parameters that contribute to non-viral gene delivery are chemical structure and charge distribution. Indeed, saccharide units have been reported to have specific interactions with proteins located in the outer leaflet of the plasma cell membrane that facilitate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service