Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A406

Sigma-Aldrich

1-Acenaphthenol

99%

Synonym(s):

1-Hydroxyacenaphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O
CAS Number:
Molecular Weight:
170.21
Beilstein:
2048222
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

145-148 °C (lit.)

SMILES string

OC1Cc2cccc3cccc1c23

InChI

1S/C12H10O/c13-11-7-9-5-1-3-8-4-2-6-10(11)12(8)9/h1-6,11,13H,7H2

InChI key

MXUCIEHYJYRTLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica R Murray et al.
Chemical research in toxicology, 32(12), 2538-2551 (2019-11-21)
3-Nitrobenzanthrone (3-NBA) is a suspected human carcinogen present in diesel exhaust. It requires metabolic activation via nitroreduction in order to form DNA adducts and promote mutagenesis. We have determined that human aldo-keto reductases (AKR1C1-1C3) and NAD(P)H:quinone oxidoreductase 1 (NQO1) contribute
Somnath Mallick
Chemosphere, 219, 748-755 (2018-12-18)
A gram-negative bacterium designated as RTSB was isolated from a petroleum-contaminated soil competent of utilizing acenaphthene as the solitary source of carbon and energy. The strain RTSB was identified as a Sphingobacterium species based on the morphological, nutritional and biochemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service