Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A36301

Sigma-Aldrich

Allyltriethoxysilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2Si(OC2H5)3
CAS Number:
Molecular Weight:
204.34
Beilstein:
1772306
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.406 (lit.)

bp

78 °C/21 mmHg (lit.)

density

0.903 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](CC=C)(OCC)OCC

InChI

1S/C9H20O3Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5H,1,6-9H2,2-4H3

InChI key

UMFJXASDGBJDEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allyltriethoxysilane is an allylsilane that is mainly used as an organo-metallic reagent. It is a low toxic reagent that can be used to modify the surface of the nanoparticles for a variety of applications which include dental composites.[1][2]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoparticle-reinforced resin-based dental composites
Xia Y, et al.
Journal of Dentistry, 36(6), 450-455 (2008)
Qihui Shen et al.
Nanomaterials (Basel, Switzerland), 9(5) (2019-05-09)
The nonspecific adsorption of proteins and bacteria on the surface of polydimethylsiloxane (PDMS) had been a serious concern in a wide range of applications, such as medical devices. In order to improve the anti-adhesive and antibacterial capability, bare silver nanoparticles
A general catalytic allylation using allyltrimethoxysilane
Yamasaki S, et al.
Journal of the American Chemical Society, 124(23), 6536-6537 (2002)
Yueer Yan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12161-12170 (2015-07-16)
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service