Skip to Content
MilliporeSigma
All Photos(1)

Documents

A22904

Sigma-Aldrich

N-Acetyl-L-tyrosine ethyl ester monohydrate

99%

Synonym(s):

ATEE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NHCOCH3)CO2C2H5·H2O
CAS Number:
Molecular Weight:
269.29
Beilstein:
2217900
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

optical activity

[α]20/D +24.1°, c = 1 in ethanol

mp

80-81 °C (lit.)

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service