Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A14405

Sigma-Aldrich

1-Acetyl-1-cyclohexene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9COCH3
CAS Number:
Molecular Weight:
124.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.49 (lit.)

bp

201-202 °C (lit.)

density

0.966 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C1=CCCCC1

InChI

1S/C8H12O/c1-7(9)8-5-3-2-4-6-8/h5H,2-4,6H2,1H3

InChI key

LTYLUDGDHUEBGX-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcelina Mazur et al.
Biomolecules, 10(1) (2020-01-16)
Starting from 1-acetyl-1-cyclohexene, three enantiomeric pairs (ee ≥99%) of bicyclic δ-halo-γ-lactones with cyclohexane ring were obtained in five-step synthesis. The key stereochemical steps were lipase-catalyzed kinetic resolution of racemic 1-(cyclohex-1-en-1-yl) ethanol followed by transfer of chirality to ethyl 2-(2-ethylidenecyclohexyl) acetate
Gilles Brackman et al.
PloS one, 6(1), e16084-e16084 (2011-01-21)
Many bacteria, including Vibrio spp., regulate virulence gene expression in a cell-density dependent way through a communication process termed quorum sensing (QS). Hence, interfering with QS could be a valuable novel antipathogenic strategy. Cinnamaldehyde has previously been shown to inhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service