Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A12658

Sigma-Aldrich

N-Acetylanthranilic acid

99%

Synonym(s):

2-Acetamidobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CONH)C6H4CO2H
CAS Number:
Molecular Weight:
179.17
Beilstein:
880371
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

off-white to tan

mp

184-187 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(=O)Nc1ccccc1C(O)=O

InChI

1S/C9H9NO3/c1-6(11)10-8-5-3-2-4-7(8)9(12)13/h2-5H,1H3,(H,10,11)(H,12,13)

InChI key

QSACCXVHEVWNMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H K Hund et al.
Biological chemistry Hoppe-Seyler, 371(10), 1005-1008 (1990-10-01)
Quinaldine catabolism was investigated with the bacterial strain Arthrobacter sp., which is able to grow aerobically in a mineral salt medium with quinaldine as sole source of carbon, nitrogen and energy. The following degradation products of quinaldine were isolated from
Nikolai E Polyakov et al.
Organic & biomolecular chemistry, 3(5), 881-885 (2005-02-26)
CIDNP techniques were applied to the investigation of the elementary mechanism of photoinduced interaction between anti-arrhythmic drug lappaconitine and amino acids tyrosine and tryptophan. It has been shown that the reactions involve the formation of lappaconitine radical anion. Lappaconitine radical
Stephan Kolkenbrock et al.
Journal of bacteriology, 188(24), 8430-8440 (2006-10-17)
N-acetylanthranilate amidase (Amq), a 32.8-kDa monomeric amide hydrolase, is involved in quinaldine degradation by Arthrobacter nitroguajacolicus Rü61a. Sequence analysis and secondary structure predictions indicated that Amq is related to carboxylesterases and belongs to the alpha/beta-hydrolase-fold superfamily of enzymes; inactivation of
Jörg Overhage et al.
Microbiology (Reading, England), 151(Pt 2), 491-500 (2005-02-09)
Arthrobacter nitroguajacolicus Rü61a, which utilizes quinaldine as sole source of carbon and energy, was shown to contain a conjugative linear plasmid of approximately 110 kb, named pAL1. It exhibits similarities with other linear plasmids from Actinomycetales in that it has
Christine Müller et al.
Applied and environmental microbiology, 80(23), 7266-7274 (2014-09-23)
A bacterial strain, which based on the sequences of its 16S rRNA, gyrB, catA, and qsdA genes, was identified as a Rhodococcus sp. closely related to Rhodococcus erythropolis, was isolated from soil by enrichment on the Pseudomonas quinolone signal [PQS;

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service