Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A10604

Sigma-Aldrich

2,5-Hexanedione

97%

Synonym(s):

Acetonylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2COCH3
CAS Number:
Molecular Weight:
114.14
Beilstein:
506525
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

0.43 mmHg ( 20 °C)

Assay

97%

refractive index

n20/D 1.425 (lit.)

bp

191 °C (lit.)

mp

−6-−5 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CCC(C)=O

InChI

1S/C6H10O2/c1-5(7)3-4-6(2)8/h3-4H2,1-2H3

InChI key

OJVAMHKKJGICOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah N Campion et al.
Toxicological sciences : an official journal of the Society of Toxicology, 117(2), 457-465 (2010-07-10)
Sertoli cells are essential for testicular germ cell maintenance and survival. We made the unexpected observation that x-radiation (x-ray)-induced germ cell loss is attenuated by co-exposure with the Sertoli cell toxicant 2,5-hexanedione (HD). The mechanisms underlying this attenuation of germ
Antônio Felipe F Oliveira et al.
Journal of analytical toxicology, 33(4), 223-228 (2009-05-28)
2,5-Hexanedione (2,5-HD) is the most important metabolite of n-hexane and methyl ethyl ketone in human urine. Urinary 2,5-HD is used as a biomarker for biological monitoring of workers exposed to n-hexane. A simple method using headspace solid-phase microextraction (HS-SPME) and
Marion Müller et al.
Organic & biomolecular chemistry, 8(7), 1540-1550 (2010-03-20)
The enantiopure (2S,5S)-hexanediol serves as a versatile building block for the production of various fine chemicals and pharmaceuticals. For industrial and commercial scale, the diol is currently obtained through bakers' yeast-mediated reduction of 2,5-hexanedione. However, this process suffers from its
Yan Sun et al.
Archives of toxicology, 86(2), 205-215 (2011-09-09)
Studies have shown that 2,5-hexanedione (2,5-HD) is the main active metabolite of n-hexane in the human body. The toxicity of n-hexane and 2,5-hexanedione has been extensively researched, but toxicity to the reproductive system, especially the impact on female reproductive function
Min-Sun Kim et al.
Toxicology, 260(1-3), 97-103 (2009-05-26)
2,5-Hexanedione (HD), a metabolite of n-hexane, causes central and peripheral neuropathy leading to motor neuron deficits. Although chronic exposure to n-hexane is known to cause gradual sensorimotor neuropathy, there are no reports on the effects of low doses of HD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service