Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0786

Sigma-Aldrich

Adipic semialdehyde methyl ester

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95%

density

1 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

COC(=O)CCCCC=O

InChI

1S/C7H12O3/c1-10-7(9)5-3-2-4-6-8/h6H,2-5H2,1H3

InChI key

FDNFXHCDOASWAY-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Bernotas et al.
The Biochemical journal, 270(2), 539-540 (1990-09-01)
An improved, high-yield synthesis of the corresponding N-carboxypentyl derivatives of three iminoalditol glycosidase inhibitors has been developed for affinity chromatography enzyme purification. Reductive amination of 1-deoxynojirimycin (or its D-manno or D-galacto analogues) with methyl 5-formylvalerate and NaBH3CN at neutral pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service