Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96837

Sigma-Aldrich

Z-DL-Lys(Z)-OH

≥98.0% (TLC)

Synonym(s):

Nα,Nε-Di-Z-DL-lysine, Nα,Nε-di-Z-DL-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O6
CAS Number:
Molecular Weight:
414.45
Beilstein:
2027663
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥98.0% (TLC)

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

90-95 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)C(CCCCNC(=O)OCc1ccccc1)NC(=O)OCc2ccccc2

InChI

1S/C22H26N2O6/c25-20(26)19(24-22(28)30-16-18-11-5-2-6-12-18)13-7-8-14-23-21(27)29-15-17-9-3-1-4-10-17/h1-6,9-12,19H,7-8,13-16H2,(H,23,27)(H,24,28)(H,25,26)

InChI key

BLZXFNUZFTZCFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service