Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96744

Sigma-Aldrich

Fmoc-Homoser(Trt)-OH

≥98.0% (HPLC)

Synonym(s):

Nα-Fmoc-O-trityl-L-homoserine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H33NO5
CAS Number:
Molecular Weight:
583.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (HPLC)

form

crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

impurities

may contain ≤15 wt. % Ethanol

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](CCOC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)OCC4c5ccccc5-c6ccccc46

InChI

1S/C38H33NO5/c40-36(41)35(39-37(42)43-26-34-32-22-12-10-20-30(32)31-21-11-13-23-33(31)34)24-25-44-38(27-14-4-1-5-15-27,28-16-6-2-7-17-28)29-18-8-3-9-19-29/h1-23,34-35H,24-26H2,(H,39,42)(H,40,41)/t35-/m0/s1

InChI key

QUTREFXHXPNEJN-DHUJRADRSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service