Skip to Content
MilliporeSigma
All Photos(1)

Documents

96170

Sigma-Aldrich

Z-Gly-NH2

≥99.0% (HPLC)

Synonym(s):

Z-glycinamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OOCNHCH2CONH2
CAS Number:
Molecular Weight:
208.21
Beilstein:
2054564
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

136-139 °C

application(s)

peptide synthesis

SMILES string

NC(=O)CNC(=O)OCc1ccccc1

InChI

1S/C10H12N2O3/c11-9(13)6-12-10(14)15-7-8-4-2-1-3-5-8/h1-5H,6-7H2,(H2,11,13)(H,12,14)

InChI key

HQYMUNCIMNFLDT-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service