Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96091

Sigma-Aldrich

N-Z-1,5-pentanediamine hydrochloride

≥98.0%

Synonym(s):

N-Z-1,5-diaminopentane hydrochloride, Benzyl N-(5-aminopentyl)carbamate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCONH(CH2)5NH2 · HCl
CAS Number:
Molecular Weight:
272.77
Beilstein:
3737595
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form:
solid
Assay:
≥98.0%

Assay

≥98.0%

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

172-175 °C

functional group

Cbz
amine

SMILES string

NCCCCCNC(OCC1=CC=CC=C1)=O.[H]Cl

InChI

1S/C13H20N2O2.ClH/c14-9-5-2-6-10-15-13(16)17-11-12-7-3-1-4-8-12;/h1,3-4,7-8H,2,5-6,9-11,14H2,(H,15,16);1H

InChI key

VYIRBXGDTOPWSY-UHFFFAOYSA-N

Other Notes

Introduction of a pentamethylene linker; Synthesis of a polyamine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.K. Choi et al.
Tetrahedron, 49, 5777-5777 (1993)
A Delbarre et al.
Journal of medicinal chemistry, 28(6), 752-761 (1985-06-01)
In order to obtain breast tumor directed agents, we have prepared mixed compounds using estradiol or (E)-clomiphene as specific vectors of the breast tissue and a DNA intercalator from the ellipticine series as the cytotoxic agent. Among the newly synthesized
P Laugâa et al.
Biochemistry, 24(20), 5567-5575 (1985-09-24)
A trimer made up of three acridine chromophores linked by a positively charged poly(aminoalkyl) chain was synthesized as a potential tris-intercalating agent. The length of the linking chain was selected to allow intercalation of each chromophore according to the excluded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service